Stampa…

Anti-Vascular Endothelial Growth Factor, Endocrine-gland-Derived Goat Polyclonal Antibody (Biotin)

Fornitore: US Biological
V2110-06E-25 V2110-06E-50
USBIV2110-06E-25EA 324 EUR
USBIV2110-06E-25 USBIV2110-06E-50
Anti-Vascular Endothelial Growth Factor, Endocrine-gland-Derived Goat Polyclonal Antibody (Biotin)
Anticorpi
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml

Type: Primary
Antigen: Vascular Endothelial Growth Factor, Endocrine-gland-Derived
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human,
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR