Stampa…
Bottles, wide neck, PTFE, with screw cap
Fornitore: Thermo Scientific
38111.KT 38112.KT 38113.KT 38114.KT 38115.KT
38111.KTEA 186 EUR
38111.KT 38112.KT 38113.KT 38114.KT 38115.KT
Bottles, wide neck, PTFE, with screw cap
Bottiglie Bottiglie in plastica
PTFE, natural, with screw cap.
  • Inert
  • Heat resistant up to +280 °C
Ideal for storage and transport of valuable or aggressive materials. It features heavy wall construction with super smooth internal finish.