Stampa…
Contact plates (Rodac) in peelable blister pack
Fornitore: LIOFILCHEM
15325 15328 15329 15332 15334 15365 15376 15360 15205
LIOF15325EA 61.4 EUR
LIOF15325 LIOF15328 LIOF15329 LIOF15332 LIOF15334 LIOF15365 LIOF15376 LIOF15360 LIOF15205
Contact plates (Rodac) in peelable blister pack
Terreni per microbiologia Piastre contact per microbiologia
The new peelable blister pack fulfills the highest quality and protection standards for the plate. The blister pack, produced fully automatically and in controlled environment, offers the following advantages compared to the pouch pack:
  • Allows the usage of single plates, leaving the remaining plates protected in their pack
  • Drastically reduces the dehydration of culture medium during the shelf life period
  • Permits better product preservation and isolation during transportation and storage
  • Guarantees the product cannot be handled after the packaging
  • Avoids contamination of the controlled environment