Stampa…
ESD safety ankle boots, lace-up, uvex motion style, 6979
Fornitore: UVEX
6979.8/43 6979.8/36 6979.8/37 6979.8/42 6979.8/49 6979.8/50 6979.8/44 6979.8/45 6979.8/46 6979.8/47 6979.8/48 6979.8/40 6979.8/38 6979.8/39 6979.8/41
114-5494EA 136 EUR
114-5494 114-5487 114-5488 114-5493 114-5533 114-5534 114-5495 114-5496 114-5497 114-5531 114-5532 114-5491 114-5489 114-5490 114-5492
ESD safety ankle boots, lace-up, uvex motion style, 6979
Scarpe e stivali
Ultralight ESD safety ankle boot made from breathable high-tech microvelour, soft padded collar and tongue, breathable distance mesh and an extremely flexible dual density PUR sole (slip-, abrasion- and cut-resistant, resistant to oil and petrol and withstands brief exposure to heat of approx. +120 °C). Free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-twist heel cap for better stability and protection from ankle twisting
  • uvex 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Virtually seamless design to eliminate the potential for pressure points and blisters
  • Elastic laces for quick and individual adjustment
  • Protective steel toecap (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black/grey
Certificazioni: EN 61340 (ESD); EN ISO 20345:2011; DGUV 112-191; EN ISO 20345:2011; S2, SRC