Stampa…

Anti-NLRC4 Rabbit Polyclonal Antibody

Fornitore: Novus Biologicals
NBP1-78979 NBP1-78979SS
NOVUNBP1-78979EA 542 EUR
NOVUNBP1-78979 NOVUNBP1-78979SS
Anti-NLRC4 Rabbit Polyclonal Antibody
Anticorpi
The CARD12 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CARD12. This antibody reacts with human, mouse. The CARD12 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: CARD12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR