Stampa…

Anti-CD36 Rabbit Polyclonal Antibody

Fornitore: Novus Biologicals
NB400-145 NB400-145SS
NOVUNB400-145EA 638 EUR
NOVUNB400-145 NOVUNB400-145SS
Anti-CD36 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR