Stampa…

Anti-PTPN5 Mouse Monoclonal Antibody [clone: 23E5]

Fornitore: Novus Biologicals
NB300-202 NB300-202SS
NOVUNB300-202EA 591 EUR
NOVUNB300-202 NOVUNB300-202SS
Anti-PTPN5 Mouse Monoclonal Antibody [clone: 23E5]
Anticorpi
Type: Primary
Antigen: PTPN5 (protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched))
Clonality: monoclonal
Clone: 2300000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR