Stampa…

Anti-HIST3H3 Rabbit Polyclonal Antibody

Fornitore: Novus Biologicals
NB21-1002 NB21-1002SS
NOVUNB21-1002EA 630 EUR
NOVUNB21-1002 NOVUNB21-1002SS
Anti-HIST3H3 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR