Stampa…
Anti-RAD50 Rabbit Polyclonal Antibody
Fornitore: Novus Biologicals
NB100-154 NB100-154SS
NOVUNB100-154EA 624 EUR
NOVUNB100-154 NOVUNB100-154SS
Anti-RAD50 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: RAD50 (RAD50 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici