Stampa…

Anti-PKHD1 Rabbit Polyclonal Antibody

Fornitore: Novus Biologicals
BC110-60479 BC110-60479SS
NOVUBC110-60479EA 591 EUR
NOVUBC110-60479 NOVUBC110-60479SS
Anti-PKHD1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: PKHD1 (polycystic kidney and hepatic disease 1 (autosomal recessive))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR