Stampa…

Anti-IgG Donkey Polyclonal Antibody (CF™ 543)

Fornitore: Biotium
20318 20318-1
20318.EA 359 EUR
20318. 20318-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 543)
Anticorpi
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 543 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR