Stampa…

Anti-IgG Goat Polyclonal Antibody (CF™ 555)

Fornitore: Biotium
20233 20233-1
20233.EA 359 EUR
20233. 20233-1
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Anticorpi
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR