Stampa…
Anti-Biotin Mouse Monoclonal Antibody (CF®640R)
Fornitore: Biotium
20207 20207-1
20207.EA 359 EUR
20207. 20207-1
Anti-Biotin Mouse Monoclonal Antibody (CF®640R)
Anticorpi
Type: Tag
Antigen: Biotin (biotin)
Clonality: monoclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Visualizza i codici