Stampa…

Anti-Biotin Mouse Monoclonal Antibody (CF®633)

Fornitore: Biotium
20206 20206-1
20206.EA 359 EUR
20206. 20206-1
Anti-Biotin Mouse Monoclonal Antibody (CF®633)
Anticorpi
Type: Tag
Antigen: Biotin (biotin)
Clonality: monoclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR