Stampa…

Anti-IgG Goat Polyclonal Antibody (CF™ 350)

Fornitore: Biotium
20198 20198-1
20198.EA 300 EUR
20198. 20198-1
Anti-IgG Goat Polyclonal Antibody (CF™ 350)
Anticorpi
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 350 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Guinea pig
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR