Stampa…

Anti-IgG Rabbit Polyclonal Antibody (CF™ 568)

Fornitore: Biotium
20093 20093-1
20093.EA 359 EUR
20093. 20093-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 568)
Anticorpi
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR