Stampa…
Anti-IgG Rabbit Polyclonal Antibody (CF™ 640R)
Fornitore: Biotium
20090 20090-1
20090.EA 300 EUR
20090. 20090-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 640R)
Anticorpi
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Goat