Stampa…

Anti-Adenosine phosphate Rabbit Monoclonal Antibody [clone: EPR8471]

Fornitore: Abcam
AB140604-100 AB140604-1ML
ABCAAB140604-100EA 663 EUR
ABCAAB140604-100 ABCAAB140604-1ML
Anti-Adenosine phosphate Rabbit Monoclonal Antibody [clone: EPR8471]
Anticorpi
Rabbit monoclonal [EPR8471] to Adenosine phosphate.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Adenosine phosphate
Clonality: Monoclonal
Clone: EPR8471
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR