Stampa…

Anti-IL-10 Rabbit Monoclonal Antibody [clone: EPR1114]

Fornitore: Abcam
AB133575-100 AB133575-1ML
ABCAAB133575-100EA 807 EUR
ABCAAB133575-100 ABCAAB133575-1ML
Anti-IL-10 Rabbit Monoclonal Antibody [clone: EPR1114]
Anticorpi
Rabbit monoclonal [EPR1114] to IL-10.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: EPR1114
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR