Stampa…
Anti-Cyclin B1 Rabbit Monoclonal Antibody [clone: Y106]
Fornitore: Abcam
AB32053-100 AB32053-1ML
ABCAAB32053-100EA 800 EUR
ABCAAB32053-100 ABCAAB32053-1ML
Anti-Cyclin B1 Rabbit Monoclonal Antibody [clone: Y106]
Anticorpi
Rabbit monoclonal [Y106] to Cyclin B1.
Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Cyclin B1
Clonality: Monoclonal
Clone: Y106
Conjugation:
Epitope: ab32053 reacts with an epitope located in teh C terminal region of Cyclin B1.
Host: Rabbit
Isotype: IgG
Reactivity: Human