Stampa…

Anti-U2AF1 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB129760-100 ORB129760-200
BIRBORB129760-100EA 0 EUR
BIRBORB129760-100 BIRBORB129760-200
Anti-U2AF1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: U2AF1 (U2 small nuclear RNA auxiliary factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR