Stampa…

Anti-ZEB2 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB128931-100 ORB128931-200
BIRBORB128931-100EA 0 EUR
BIRBORB128931-100 BIRBORB128931-200
Anti-ZEB2 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: ZEB2 (zinc finger E-box binding homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR