Stampa…

Anti-Sodium Channel-pan Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB128704-100 ORB128704-200
BIRBORB128704-100EA 0 EUR
BIRBORB128704-100 BIRBORB128704-200
Anti-Sodium Channel-pan Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Sodium Channel-pan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR