Stampa…

Anti-IR Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB128223-100 ORB128223-200
BIRBORB128223-100EA 0 EUR
BIRBORB128223-100 BIRBORB128223-200
Anti-IR Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: IR
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR