Stampa…
Anti-HOXA1/B1/D1 Rabbit Polyclonal Antibody
Fornitore: Biorbyt
ORB126984-100 ORB126984-200
BIRBORB126984-100EA 0 EUR
BIRBORB126984-100 BIRBORB126984-200
Anti-HOXA1/B1/D1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: HOXA1/B1/D1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici