Stampa…

Anti-p18 INK Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB126801-100 ORB126801-200
BIRBORB126801-100EA 0 EUR
BIRBORB126801-100 BIRBORB126801-200
Anti-p18 INK Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: p18 INK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR