Stampa…

Anti-PHLA2 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB126247-100 ORB126247-200
BIRBORB126247-100EA 0 EUR
BIRBORB126247-100 BIRBORB126247-200
Anti-PHLA2 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: PHLA2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR