Stampa…

Anti-ARHGEF2 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB125448-100 ORB125448-200
BIRBORB125448-100EA 0 EUR
BIRBORB125448-100 BIRBORB125448-200
Anti-ARHGEF2 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: ARHGEF2 (Rho/Rac guanine nucleotide exchange factor (GEF) 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR