Stampa…

Anti-IR Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB106334-100 ORB106334-20
BIRBORB106334-100EA 0 EUR
BIRBORB106334-100 BIRBORB106334-20
Anti-IR Rabbit Polyclonal Antibody
Anticorpi
Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: IR
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR