Stampa…

Anti-CCNB1IP1 Rabbit polyclonal antibody

Fornitore: Biorbyt
ORB100825-200 ORB100825-100
BIRBORB100825-200EA 0 EUR
BIRBORB100825-200 BIRBORB100825-100
Anti-CCNB1IP1 Rabbit polyclonal antibody
Anticorpi
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: CCNB1IP1 (cyclin B1 interacting protein 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR