Stampa…

Anti-LITAF Mouse Monoclonal Antibody

Fornitore: Biorbyt
ORB22615-100 ORB22615-20 ORB22615-5
BIRBORB22615-100EA 0 EUR
BIRBORB22615-100 BIRBORB22615-20 BIRBORB22615-5
Anti-LITAF Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: LITAF (lipopolysaccharide-induced TNF factor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR