Stampa…

Anti-AMPH Goat Polyclonal Antibody

Fornitore: Biorbyt
ORB19251-100 ORB19251-20
BIRBORB19251-100EA 0 EUR
BIRBORB19251-100 BIRBORB19251-20
Anti-AMPH Goat Polyclonal Antibody
Anticorpi
ELISA: 1:16000; Western Blot: 0.1-0.5 ?g/ml

Type: Primary
Antigen: AMPH (amphiphysin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR