Stampa…

Anti-ROS1 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB13678-100 ORB13678-200
BIRBORB13678-100EA 0 EUR
BIRBORB13678-100 BIRBORB13678-200
Anti-ROS1 Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500

Type: Primary
Antigen: ROS1 (c-ros oncogene 1 , receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR