Stampa…
Anti-ITGA5/B1 Rabbit Polyclonal Antibody
Fornitore: Biorbyt
ORB13515-100 ORB13515-20 ORB13515-200 ORB13515-5
BIRBORB13515-100EA 0 EUR
BIRBORB13515-100 BIRBORB13515-20 BIRBORB13515-200 BIRBORB13515-5
Anti-ITGA5/B1 Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: ITGA5/B1 (integrin, alpha 5/beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici