Stampa…

Anti-Envelope Glycoprotein Yellow Fever Virus Rabbit polyclonal antibody

Fornitore: Biorbyt
ORB13392-100 ORB13392-200
BIRBORB13392-100EA 0 EUR
BIRBORB13392-100 BIRBORB13392-200
Anti-Envelope Glycoprotein Yellow Fever Virus Rabbit polyclonal antibody
Anticorpi
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Envelope Glycoprotein Yellow Fever Virus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR