Stampa…

Anti-CD45RO Mouse Monoclonal Antibody (PE (Phycoerythrin))

Fornitore: Biorbyt
ORB12074-100 ORB12074-50
BIRBORB12074-100EA 0 EUR
BIRBORB12074-100 BIRBORB12074-50
Anti-CD45RO Mouse Monoclonal Antibody (PE (Phycoerythrin))
Anticorpi
Type: Primary
Antigen: CD45RO (CD45RO)
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR