Stampa…

Anti-CD45RA Mouse Monoclonal Antibody (Biotin)

Fornitore: Biorbyt
ORB12071-100 ORB12071-500
BIRBORB12071-100EA 0 EUR
BIRBORB12071-100 BIRBORB12071-500
Anti-CD45RA Mouse Monoclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: CD45RA (CD45RA)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR