Stampa…

Anti-FCGR3A Mouse Monoclonal Antibody (Biotin)

Fornitore: Biorbyt
ORB11985-100 ORB11985-500
BIRBORB11985-100EA 0 EUR
BIRBORB11985-100 BIRBORB11985-500
Anti-FCGR3A Mouse Monoclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: FCGR3A (Fc fragment of IgG, low affinity IIIa, receptor (CD16a))
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR