Stampa…

Anti-ITGA3/B1 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB10925-100 ORB10925-20 ORB10925-200 ORB10925-5
BIRBORB10925-100EA 0 EUR
BIRBORB10925-100 BIRBORB10925-20 BIRBORB10925-200 BIRBORB10925-5
Anti-ITGA3/B1 Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: ITGA3/B1 (integrin, alpha 3/beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR