Stampa…

Anti-HSPA9 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB10746-100 ORB10746-200
BIRBORB10746-100EA 0 EUR
BIRBORB10746-100 BIRBORB10746-200
Anti-HSPA9 Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: HSPA9 (heat shock 70kDa protein 9 (mortalin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR