Stampa…

Anti-Gentamicin Rabbit polyclonal antibody

Fornitore: Biorbyt
ORB10384-100 ORB10384-200
BIRBORB10384-100EA 0 EUR
BIRBORB10384-100 BIRBORB10384-200
Anti-Gentamicin Rabbit polyclonal antibody
Anticorpi
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Gentamicin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR