Stampa…

Anti-SPHK1 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB7002-100 ORB7002-200
BIRBORB7002-100EA 0 EUR
BIRBORB7002-100 BIRBORB7002-200
Anti-SPHK1 Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: SPHK1 (sphingosine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR