Stampa…
Anti-MDH1 Rabbit Polyclonal Antibody
Fornitore: Biorbyt
ORB6403-100 ORB6403-200
BIRBORB6403-100EA 0 EUR
BIRBORB6403-100 BIRBORB6403-200
Anti-MDH1 Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: MDH1 (malate dehydrogenase 1, NAD (soluble))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici