Stampa…

Anti-IL18 Receptor Beta Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB5545-100 ORB5545-200
BIRBORB5545-100EA 0 EUR
BIRBORB5545-100 BIRBORB5545-200
Anti-IL18 Receptor Beta Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: IL18 Receptor Beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR