Stampa…
Anti-SQLE Rabbit Polyclonal Antibody
Fornitore: Biorbyt
ORB4562-100 ORB4562-200
BIRBORB4562-100EA 0 EUR
BIRBORB4562-100 BIRBORB4562-200
Anti-SQLE Rabbit Polyclonal Antibody
Anticorpi
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: SQLE (squalene epoxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici