Stampa…

Anti-GFAP delta Rabbit polyclonal antibody

Fornitore: Biorbyt
ORB2836-100 ORB2836-200
BIRBORB2836-100EA 0 EUR
BIRBORB2836-100 BIRBORB2836-200
Anti-GFAP delta Rabbit polyclonal antibody
Anticorpi
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: GFAP delta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR