Stampa…

Anti-MROH9 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB2031-100 ORB2031-200
BIRBORB2031-100EA 0 EUR
BIRBORB2031-100 BIRBORB2031-200
Anti-MROH9 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: MROH9 (maestro heat-like repeat family member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR