Stampa…

Anti-TMEM147 Rabbit Polyclonal Antibody

Fornitore: Biorbyt
ORB1230-100 ORB1230-200
BIRBORB1230-100EA 0 EUR
BIRBORB1230-100 BIRBORB1230-200
Anti-TMEM147 Rabbit Polyclonal Antibody
Anticorpi
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: TMEM147 (transmembrane protein 147)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR