Stampa…

Anti-TER-119 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: TER-119]

Fornitore: Tonbo Biosciences
20-5921-U100 20-5921-U025
20-5921-U100EA 120 EUR
20-5921-U100 20-5921-U025
Anti-TER-119 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: TER-119]
Anticorpi
The TER-119 antibody is named for the antigen to which it binds, a 52 kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.

Type: Primary
Antigen: TER-119 (TER-119)
Clonality: monoclonal
Clone: TER-119
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR