Stampa…
Rotary evaporators, Hei-VAP Value Digital
Fornitore: Heidolph Instruments GmbH & Co.KG
560-01102-03 560-01102-04 560-01112-03 560-01112-04 560-01302-03 560-01302-04 560-01312-03 560-01312-04 560-01502-03 560-01502-04 560-01512-03 560-01512-04 560-01602-03 560-01602-04 560-01612-03 560-01612-04 560-01307-04 560-01307-03 560-01317-04 560-01317-03
HEID560-01102-03EA 3060 EUR
HEID560-01102-03 HEID560-01102-04 HEID560-01112-03 HEID560-01112-04 HEID560-01302-03 HEID560-01302-04 HEID560-01312-03 HEID560-01312-04 HEID560-01502-03 HEID560-01502-04 HEID560-01512-03 HEID560-01512-04 HEID560-01602-03 HEID560-01602-04 HEID560-01612-03 HEID560-01612-04 HEID560-01307-04 HEID560-01307-03 HEID560-01317-04 HEID560-01317-03
Rotary evaporators, Hei-VAP Value Digital
Evaporatori Evaporatori rotanti
These Hei-VAP devices with hand lift are optimally suited for all standard applications. Parameters can be controlled with the simple turn of a knob.
  • With digital display for set and actual heating bath temperature
  • Large dial controls for adjustment of rotation speed and bath temperature
  • Required angle and height of the evaporating flask can adjusted individually
  • Patented flask clamp Easy-Clip allows a simple and safe removing of the evaporating flask due to the force of unscrewing the Easy-Clip
  • Detachable operating panel enables safe and convenient handling from outside closed fume hoods
  • Separate on/off switch for heating prevents unintentional heat-up – the button is illuminated for visual control
  • FDA-compliant, transparent and uv-resistant tubing, with higher chemical resistance
210 °C safety heating bath with handles and pour spout will power off if temperature overshoots by 5 °C or runs dry. The bath power cable coupling complies with protection class IP 67 to prevent corrosion and short circuits.
Glassware set G1
Diagonal condenser for all standard distillations; the option with the most affordable price.
Glassware set G3
Vertical condenser for all standard distillations; the option with the smallest space requirements, also available as XL version (condensing surface of 2200 cm² for up to 40% higher evaporation rates)
Glassware set G5
Dry Ice condenser for low-boiling solvents.
Glassware set G6
Vertical condenser; centerpiece with valve for reflux distillation.
Informazioni per l'ordine: Supplied with water/oil bath, 1 L evaporation flask (29/32), 1 L receiving flask (35/20) and condenser glassware set.
Please note: A vacuum source, vacuum check valve and cooling system are required but not supplied. Please contact your local Avantor sales office for advice and details.
Attenzione: * con plastica trasparente